Learn More
MSA™ Flexi-Filter™ Pads for Advantage™ Respirators
Manufacturer:  MSA™ 818342
 View more versions of this product
Catalog No. 19-039-693
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Advantage™ Flexi-Filters™ Pad
Magenta
EN 143, NIOSH, USA
P100 Filter
99.97%
Bayonet
Asbestos abatement, painting, sanding, grinding, welding
Advantage™ respirator
2/Pk.
Filter pad only