MSA™ Flexi-Filter™ Pads for Advantage™ Respirators

Manufacturer: MSA™ 818355

 View more versions of this product

Catalog No. 19-052-713

  Check Availability
Add to Cart

    Specifications

    Quantity 2 Pack
    For Use With (Equipment) Advantage™ respirator,Advantage™ respirator
    Product Type Advantage™ Flexi-Filters™ Pad
    For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
    Includes Nuisance level OV removal
    Certifications EN 143, NIOSH, USA
    Design Type Swept back
    Filter Efficiency 95%
    Filter Type P95 Filter
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit