Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA™ FP Pro™ Vest Style Harnesses
Easy to doff and don, this harness features a full-body design offering excellent strength
Supplier: MSA™ 10033839
View more versions of this product
Safety Services Available
Overview
Fall Protection Assessment
Description
- Provides comfortable protection after long use in extreme conditions due to lightweight features and durable webbing
- Integral belt and back pad for additional comfort and support for work positioning applications
- Fits wide range of body types and sizes
- Color-contrasting thigh and torso straps simplify donning
- Bright red polyester webbing for high visibility
- Capacity of 181.4kg (400 lb.)
Specifications
Standard | |
Tongue | |
Back, Hips |
Meets OSHA and ANSI requirements; CSA certified | |
Vest |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction