MSA™ Gravity™ Harnesses

Curvilinear Comfort System provides total comfort during use

Manufacturer: MSA™ 10067414

 View more versions of this product

Catalog No. 19-150-955

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Style Cross-Over
    Certifications/Compliance CSA certified; Certified according EN 361
    Color Red/Blue
    For Use With (Application) Ladder climbing, controlled descent, rescue, fall arrest
    Size X-Small
    D-Ring Location Back, Front, Shoulder, Side
    Material Nylon

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit