MSA™ Lynx™ Confined Space Entry Kit
Available with tripod or folding davit
Manufacturer: MSA™ 10024020
 View more versions of this product
Catalog No. 19-815-770
  Check Availability
Add to Cart
    Specifications
    Load Bearing Capacity (Metric) 281kg
    Includes Folding davit, Lynx rescuer with mounting bracket, side mount Lynx hoist, split mount pulley, carrying bags and training video
    Certifications/Compliance ANSI Z359.1, ANSI A10.14, OSHA
    For Use With (Application) Ideal for multiple confined space
    Load Bearing Capacity (English) 610 lb.
  • Description & Specifications
Related Products
Product Content Feedback