MSA™ NFL™ V-Gard™ Caps
Official NFL insignias help promote regular use on the job
Manufacturer: MSA™ 818403
 View more versions of this product
Catalog No. 19-088-1103
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance Meets or exceeds ANSI Z89.1-2003; Type I; Classes G and E.
    Color New York Giants
    Material HDPE
    Suspension Type 1-Touch
  • Description & Specifications
Related Products
Product Content Feedback