Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSA OptimAir™ TL PAPR Standard Kit
Standard kits come complete with PAPR, battery, charger, and appropriate breathing tube
Supplier: MSA 10081115
Description
- Available with facepiece or hood
- Motor blower electronics allow for optimum airflow
- Compact and ergonomic battery pack available in standard or extended-life
- Automatic flow rates
- Crushproof breathing tube and sealing gaskets on cartridges
- Audible and visual alarms for low flow indication and low battery voltage
- Suitable applications include chemical, hazardous materials, asbestos, mold and lead paint abatement, and pharmaceutical
Specifications
Lithium-ion | |
PAPR Assembly with Facepiece | |
Chemical, hazardous materials, asbestos, mold and lead paint abatement, and pharmaceutical | |
Motor blower, smart charger, standard battery, hose and decon belt | |
10081115 | |
1.8 kg |
NIOSH | |
Facepiece | |
Optimair™ tl powered air purifying respirator | |
No | |
4 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction