MSA™ MSA™ Air-Line Respirators and Air-Supplied Hood Accessory, Quick-Disconnect Hansen SS Male Plug
Manufacturer: MSA™ 628208
Catalog No. 18-999-3720
  Check Availability
Add to Cart
    For Use With (Equipment) Airline Respirator and Air-Supplied Hood
    Product Type Quick-Disconnect Hansen Male Plug
    For Use With (Application) Connect air supply hoses to manifold and air source
    Includes 0.25 in. npt female
    Material Stainless Steel
  • Description & Specifications
Related Products
Product Content Feedback