MSA™ Replacement Suspensions for V-Gard™ Caps and Hats

Manufacturer: MSA™ 492567

 View more versions of this product

Catalog No. 18-999-3982

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Size Small
    Construction 4 Point webbed suspension, adjustable dual Nylon crown straps, perforated Vinyl sweatband, large easy adjustment knob
    Includes Headband, nape strap, sweatband and adjustable crown straps
    Color White
    Material Nylon
    Product Type Fas-Trac™ Ratchet Suspension
    For Use With (Equipment) V-Gard™ Hard Hat and Cap

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit