Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ SCBA Replacement Cylinders: With Valve

Manufacturer:  MSA™ 807587

 View more versions of this product

Catalog No. 19-815-691

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

1.27cm3
SCBA
Firefighting
Carbon Wrapped Aluminum
3.3kg
45 cu. ft., 1.27cm3
45 cu. ft.
Gas Cylinder, H-30 Stealth
Valve
4500psi
7 lb.
30 min.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit