Learn More
MSA™ Sirius™ Multigas Detector Calibration Flow Control Kits
Flow control kits for multigas detectors.
Supplier: MSA™ 10050984
Description
- For use with multigas detectors
- Flowrate: 0.25 LPM
- Includes tubing and fitting less gas
Specifications
| Multigas Detector | |
| Fixed Flow Regulator Kit | |
| 0.25 LPM |