Learn More
MSA™ Sound Control SH™ Ear Muffs
Designed to fit snugly into new accessory slots on V-Gard™ slotted hard hat
Manufacturer:  MSA™ 10026398
Catalog No. 19-120-876
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Spring design for low pressure and high level of comfort
  • Three distinct wearing positions for ease of use
  • Excellent grip to adjust cups easily, even when wearing gloves
  • Provides maximum space for ears inside cup
  • Soft ear cup material for extended wear
  • Easy ear cup height adjustment
  • Retain Class E dielectric rating on the V-Gard full brim hard hat
  • Used for sanding and grinding
Compliance
Third-party certified in accordance with ANSI S3.19-1974, Meet CSA Class A
Specifications
Specifications
ANSI S3.19-1974, CSA Class A
For font brim hard caps
Cap mount
Sanding and grinding
26dB
Ear Muff