Learn More
MSA™ TechnaCurv™ Full-Body Harnesses: Pullover Style
Made of soft, flexible webbing
Manufacturer:  MSA™ 10041609
 View more versions of this product
Catalog No. 17-200-109
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pullover-style, full-body harness
Red
Pullover full body
Back; Chest; Side
Manual
ANSI Z359.1, A10.32, CSA Z259.10, OSHA
Standard
Leg strap: Tongue; Chest strap: Qwik-Fit
Polyester
Promotions
Promotions
  • Wearwell Tennessee Mat Company Inc. , Fisherbrand , MSA
    Save 30% on safety products that protect your workers, and when accidents happen, allow you to respond instantly.
    : SAFETY30
    07-14-2017