MSA™ TechnaCurv™ Full-Body Harnesses: Vest Style

Made of soft, flexible webbing

Manufacturer: MSA™ 10041595

 View more versions of this product

Catalog No. 17-200-104

  Check Availability
Add to Cart

    Specifications

    Product Type Full-body harness
    Color Red
    Style Vest
    Buckle Style Tongue
    D-Ring Location Back; Hips
    Webbing Material Teflon-coated Polyester
    Capacity (English) 400 lb.
    Includes Qwik-fit chest strap, manual
    Buckle Style (Leg) Tongue
    Certifications/Compliance OSHA, ANSI Z359.1 and A10.32, CSA Z259.10. Certified according to EN 361and EN 358
    For Use With (Application) Confined space, construction
    Size Standard

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit