Learn More
MSA™ Ultra Elite™ M7 Facepiece
For use with FireHawk™ M7 air masks
Supplier: MSA™ 10084689
View more versions of this product
Safety Services Available
Overview
Repair/Recertification
Includes: Speed-on™ head harness, nosecup, firehawk™ m7 heads up display bracket, standard speaking diaphragm
Description
Provides enhanced peripheral and downward vision
- Resists chemical attack, temperature extremes, and rugged use without tearing
- Seals securely with wide range of facial contours
- Five adjustable straps with stainless-steel, field-replaceable buckles for comfortable fit
- Standard-speaking diaphragm
- Rugged polycarbonate lens with scratch-resistant coating
- Choice of push-to-connect or slide-to-connect style regulators
Specifications
Facepiece Assembly | |
Firehawk™ m7 air mask | |
Small |
Resists chemical attack, temperature extremes, rugged use without tearing | |
Speed-on™ head harness, nosecup, Firehawk™ M7 heads-up display bracket, standard speaking diaphragm |