Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Ultravue™ Full-Facepiece Respirator: Black Facepiece Assembly

Accessories for full-facepiece respirators

Specifications

Color Black
Product Type Facepiece Assembly
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
19-076969
View Documents
MSA™
480255
OptiMair™ 6A PAPR
Each for $713.95
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-003-375
View Documents
MSA™
801173
Belt mounted scba models
Each for $1,179.20
N/A
Due to product restrictions, please to purchase or view availability for this product.
18-999-3225
View Documents
MSA™
801450
Ultralite II and custom 4500 II air mask Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

For use with MSA Ultravue Full-Facepiece Respirators

  • Model 19-003-375 for use with belt mounted SCBA models
  • Model 18-999-3225 (Small) for use with Ultralite II and Custom 4500 II air masks
  • Model 19-076-969 (Large) for use with OptimAir™ 6A PAPRs

Specifications

Specifications

Black
Facepiece Assembly
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit