MSA™ V-Gard™ Protective Slotted Cap with Staz-On™ Suspension
Lightweight for all-day wearing comfort
Manufacturer: MSA™ 815558
 View more versions of this product
Catalog No. 18-999-3991
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Size Standard
    Certifications/Compliance Meets or exceeds ANSI Z89.1-2009; Type I; Class E; SEI certified.
    Color Hi-Vis Lime Green
    Suspension Type Staz-On Pin-Lock
  • Description & Specifications
Related Products
Product Content Feedback