MSA™ V-Gard™ Replacement Polycarbonate Visors

Help protect wearers from flying debris, splash or spray

Manufacturer: MSA™ 10115840

 View more versions of this product

Catalog No. 17-200-033

  Check Availability
Add to Cart

    Specifications

    Size 8 x 17 x 0.06 in.
    For Use With (Application) Sanding and grinding, food processes, painting
    Color Clear
    Material Polycarbonate
    Certifications/Compliance EN 166, ANSI, ISEA Z87.1-2010, AS, NZS 1337. 17-200-033, CSA Z94.3
    Includes Side contours
    Product Type V-Gard™ Visor
    For Use With (Equipment) V-Gard™ Frame and Geadgear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit