MSA™ V-Gard™ Slotted Protective Cap with 1-Touch Suspension
Lightweight for all-day comfort
Manufacturer: MSA™ 10061514
 View more versions of this product
Catalog No. 19-130-3437
  Check Availability
Add to Cart
Alert
Not specifically designed to protect against lateral blows from the front, side or rear.
    Specifications
    Size Standard
    Certifications/Compliance Meets or exceeds ANSI Z89.1-2003; Type I; Class E and CSA Z94.1 2005; SEI certified
    Color Yellow; Green
    Material Polyethylene
    Suspension Type 1-Touch
  • Description & Specifications
Related Products
Product Content Feedback