MSA™ Ventilation Smoke Tube Kits
Manufacturer: MSA™ 5607
 View more versions of this product
Catalog No. 19-037-817
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Description Ventilation Smoke Kit, including aspirator bulb, six tube caps, and two glass smoke-producing tubes contained in plastic carrying case
    Quantity 1 Kit
    Tip Type Break-off
    Material Glass
    • Allows for controlled generation of visible smoke
    • Determines velocity and flow patterns of heating, ventilating and air-conditioning systems
    • Available with glass or plastic tubes with break-off tips
  • Description & Specifications
Related Products
Product Content Feedback