MSA™ Workman™ Shock-Absorbing Lanyards

Adjustable, 6 ft. (1.8m) polyester

Manufacturer: MSA™ 10072474

 View more versions of this product

Catalog No. 19-120-4004

  Check Availability
Add to Cart

    Specifications

    Adjustable Yes
    Certifications/Compliance Meets ANSI A10.32, ANSI Z359.1-1992 (R1999); and CSA Z259.11-05 E4OSHA 1910.66 and 1926.502 standards.
    Connector Types LC Snap Hook
    Length (English) 6 ft.
    Length (Metric) 1.8m

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit