Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ MSC 2032964A Inhibitor

Low molecular weight, organic compound that may help regulate biological process.

Specifications

CAS 1124381-43-6
Formula Weight 362.41g/mol
For Use With (Application) Potent and selective ASK1 inhibitor
Molecular Formula C16H13F3N6O
Storage Requirements Store at 20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB564110MG View Documents Novus Biologicals™
NB564110MG
10mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
NB564150MG View Documents Novus Biologicals™
NB564150MG
50mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Biological Activity: Potent and selective ASK1 inhibitor (IC50 = 93 nM). Selective for ASK1 over a panel of 210 kinases. Blocks LPS-induced ASK1 and p38 phosphorylation in cultured mouse astrocytes. Suppresses neuroinflammation in a mouse EAE model. Orally bioavailable and brain penetrant.

Storage: Store at 4°C.

Note: Inhibitors are guaranteed for 1 year from date of receipt.

Specifications

Specifications

1124381-43-6
Potent and selective ASK1 inhibitor
Store at 20°C
362.41g/mol
C16H13F3N6O
ASK1

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit