Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551015

Catalog No. PA551015


Add to cart

Description

Description

MSC Polyclonal Antibody detects endogenous levels of MSC protein.

Transcription repressor capable of inhibiting the transactivation capability of TCF3/E47. May play a role in regulating antigen-dependent B-cell differentiation.
Specifications

Specifications

MSC
Polyclonal
Unconjugated
MSC
Liquid
MSC
Synthesized peptide derived from the C-terminal region of human Musculin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
O60682, O88940
ABF1, BHLHA22
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
17681, 9242
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit