Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH2 Mouse anti-Human, Clone: 25D12, Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $286.00
Specifications
Antigen | MSH2 |
---|---|
Clone | 25D12 |
Dilution | Immunohistochemistry-Paraffin 1:10-1:25, Immunohistochemistry-Frozen 1:10 - 1:2000 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Monoclonal |
Description
MSH2 Monoclonal specifically detects MSH2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
MSH2 | |
Immunohistochemistry-Paraffin 1:10-1:25, Immunohistochemistry-Frozen 1:10 - 1:2000 | |
Monoclonal | |
Supernatant | |
RUO | |
Human | |
4436 | |
BALB/C mice were injected with recombinant human MSH2 protein. | |
Primary | |
Store at 4C. Do not freeze. |
25D12 | |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors | |
COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 | |
MSH2 | |
IgG1 κ | |
Tissue culture supernatant | |
This antibody reacts with a 102 kDa protein known as MSH2 (mismatch repair protein 2). |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title