Learn More
MSH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 155201AP
Catalog No. 50-173-0284
Add to cart
Description
Description
Plays a role in neurogenesis and neuronal migration By similarity. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
Specifications
Specifications
MSH2
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P43246, P43247, P54275
COCA1, FCC1, hMSH2, HNPCC, HNPCC1, LCFS2, MSH2, MutS protein homolog 2
Rabbit
IgG
150 μL
-20°C
Primary
17685, 4436, 81709
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
MSH2
Liquid
Msh2
MSH2 Fusion Protein Ag7835
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat