Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSH3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | MSH3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023260
|
Novus Biologicals
NBP294353100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023261
|
Novus Biologicals
NBP29435320UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
MSH3 Polyclonal antibody specifically detects MSH3 in Human samples. It is validated for Western BlotSpecifications
MSH3 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Divergent upstream protein, DNA mismatch repair protein Msh3, DUC1, DUG, DUPMGC163306, hMSH3, Mismatch repair protein 1, MRP1MGC163308, mutS (E. coli) homolog 3, mutS homolog 3 (E. coli) | |
Recombinant fusion protein containing a sequence corresponding to amino acids 617-761 of human MSH3 (NP_004696.2). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA Repair, Mismatch Repair, Neuroscience | |
PBS (pH 7.3), 50% glycerol | |
4437 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title