Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSH5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581222
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human MSH5.
MSH5 is a member of the mutS family of proteins that are involved in DNA mismatch repair or meiotic recombination processes. This protein is similar to a Saccharomyces cerevisiae protein that participates in meiotic segregation fidelity and crossing-over. This protein forms heterooligomers with another member of this family, mutS homolog 4. This gene encodes a member of the mutS family of proteins that are involved in DNA mismatch repair or meiotic recombination processes. This protein is similar to a Saccharomyces cerevisiae protein that participates in meiotic segregation fidelity and crossing-over. This protein forms heterooligomers with another member of this family, mutS homolog 4. Alternative splicing results in four transcript variants encoding three different isoforms.
Specifications
MSH5 | |
Polyclonal | |
PBS with no preservative | |
O43196 | |
MSH5 | |
A synthetic peptide corresponding to the center region of the human MSH5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MSH5 | |
DKFZp434C1615; G7; hMSH5; MGC2939; MSH5; Mut5; mutS homolog 5; mutS homolog 5 (E. coli); mutS protein homolog 5; MUTSH5; NG23 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
4439 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction