Learn More
MSI2 Mouse anti-Human, Clone: OTI2F10, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA506196
Catalog No. 50-167-6531
Add to cart
Description
Description
Msi2 (musashi homolog 2), also known as MSI2H, is a 328 amino acid protein that localizes to the cytoplasm and contains two RRM (RNA recognition motif) domains. Expressed ubiquitously at low levels, Msi2 functions as an RNA binding protein that, by regulating the expression of target mRNAs, is thought to play a role in the proliferation and maintece of stem cells within the central nervous system. Msi2 is subject to posttranslational phosphorylation and is upregulated in response to brain injury, suggesting a role in healing and brain tissue regeneration. Chromosomal aberrations involving the Msi2 gene are associated with the progression of chronic myeloid leukemia. Multiple isoforms of Msi2 exist due to alternative splicing events.Tissue specificity: Ubiquitous; detected at low levels.
Specifications
Specifications
MSI2
Monoclonal
Unconjugated
MSI2
Liquid
MSI2
Full length human recombit protein of human MSI2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2F10
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96DH6
MSI2H
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
124540