Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSK1/RPS6KA5 Mouse anti-Human, Clone: PCRP-RPS6KA5-1A8, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | MSK1/RPS6KA5 |
---|---|
Clone | PCRP-RPS6KA5-1A8 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122586
|
Novus Biologicals
NBP30755620UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
MSK1/RPS6KA5 Monoclonal specifically detects MSK1/RPS6KA5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
MSK1/RPS6KA5 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Phospho Specific, Protein Kinase, Signal Transduction | |
10 mM PBS with 0.05% BSA | |
9252 | |
IgG2b | |
Protein A or G purified |
PCRP-RPS6KA5-1A8 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.11, MGC1911,90 kDa ribosomal protein S6 kinase 5, MSK1EC 2.7.11.1, MSPK1, Nuclear mitogen- and stress-activated protein kinase 1, ribosomal protein S6 kinase alpha-5, ribosomal protein S6 kinase, 90kD, polypeptide 5, ribosomal protein S6 kinase, 90kDa, polypeptide 5, RLPK, RSKL, RSK-like protein kinase, S6K-alpha-5 | |
Recombinant full-length human MSK1/RPS6KA5 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title