Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548690
Description
Predicted to react with mouse based on sequence homology.
Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at 'Lys-16' (H4K16ac) which is implicated in the formation of higher-order chromatin structure (PubMed:16227571). Greatly enhances MSL2 E3 ubiquitin ligase activity, promoting monoubiquitination of histone H2B at 'Lys-34' (H2BK34Ub) (PubMed:21726816). This modification in turn stimulates histone H3 methylation at 'Lys-4' (H3K4me) and 'Lys-79' (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1 (PubMed:21726816). In the MSL complex, acts as a scaffold to tether MSL3 and KAT8 together for enzymatic activity regulation (PubMed:22547026). [UniProt].
Specifications
MSL1 | |
Polyclonal | |
Unconjugated | |
MSL1 | |
2810017F12Rik; 4121402D02Rik; 4930463F05Rik; AA682082; hampin; hMSL1; male specific lethal 1 homolog; male-specific lethal 1 homolog; male-specific lethal 1 homolog (Drosophila); male-specific lethal 1-like 1; Male-specific lethal-1 homolog 1; Msl1; Msl-1; MSL1L1; MSL1-like 1; RGD1564778 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
339287 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q68DK7 | |
MSL1 | |
KLH conjugated synthetic peptide between 339-367 amino acids from the Central region of human MSL1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction