Learn More
MSL1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 243731AP
Catalog No. 50-173-4322
Add to cart
Description
Description
Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at  ft.Lys-16 ft. (H4K16ac) which is implicated in the formation of higher-order chromatin structure. Greatly enhances MSL2 E3 ubiquitin ligase activity, promoting monoubiquitination of histone H2B at  ft.Lys-34 ft. (H2BK34Ub). This modification in turn stimulates histone H3 methylation at  ft.Lys-4 ft. (H3K4me) and  ft.Lys-79 ft. (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1. In the MSL complex, acts as a scaffold to tether MSL3 and KAT8 together for enzymatic activity regulation.
Specifications
Specifications
MSL1
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q68DK7
hMSL1, Male specific lethal 1 homolog, Male specific lethal 1 like 1, MSL 1, MSL1, MSL1 like 1, MSL1L1
Rabbit
IgG
150 μL
-20°C
Primary
339287
Western Blot
Unconjugated
MSL1
Liquid
MSL1
MSL1 Fusion Protein Ag18024
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human