Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MSLN Mouse anti-Human, Clone: OTI5B12, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF805171
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein that is present on the surface of normal mesothelium and is overexpressed in epithelial ovarian cancer and maligt mesotheliomas.Specifications
MSLN | |
Monoclonal | |
Unconjugated | |
MSLN | |
Lyophilized | |
MSLN | |
Human recombit protein fragment corresponding to amino acids 270-493 of human MSLN produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Western Blot | |
OTI5B12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13421 | |
MPF, SMRP | |
Mouse | |
IgG1 | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
10232 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title