Learn More
MSMB Mouse anti-Human, Clone: OTI5A8, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803519
Catalog No. 50-166-6550
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
MSMB protein is a member of the immunoglobulin binding factor family. It is synthesized by the epithelial cells of the prostate gland and secreted into the seminal plasma. This protein has inhibin-like activity. It may have a role as an autocrine paracrine factor in uterine, breast and other female reproductive tissues. The expression of the encoded protein is found to be decreased in prostate cancer. Two alternatively spliced transcript variants encoding different isoforms are described for this gene. The use of alternate polyadenylation sites has been found for this gene.
Specifications
Specifications
MSMB
Monoclonal
Unconjugated
MSMB
Lyophilized
MSMB
Purified prostate specific antigen (MSMB) from human seminal fluid
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5A8
PBS with 8% trehalose and no preservative; pH 7.3
P08118
HPC13, IGBF, MSP, MSPB, PN44, PRPS, PSP, PSP-94, PSP57, PSP94
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4477