Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSOP, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

66515-29-5
C4H10NO6P
199.099
GSFCOAGADOGIGE-UHFFFAOYSA-N
(RS)-α-Methylserine-O-phosphate
3964633

Specific group III mGlu antagonist

Selective group III metabotropic glutamate receptor antagonist. Displays an apparent KD?of 51 μM for the L-AP4-sensitive presynaptic mGluR on primary afferent terminals in spinal cord compared to > 700 μM for the (1S,3S)-ACPD-sensitive presynaptic mGlu in the same system. Has no activity on postsynaptic mGlu receptors or on ionotropic glutamate receptors on neonatal rat motoneurons. Also available as part of the Group III mGlu Receptor Tocriset™ .
SMILES CC(COP(=O)(O)O)(C(=O)O)N
IUPAC Name 2-amino-2-methyl-3-phosphonooxypropanoic acid
Formula Weight 199.1
Quantity 10mg
Chemical Name or Material MSOP
Grade >98%

Recommended Storage : Store at Room Temperature