Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSP1 Mouse, Clone: 6G2, Abnova™

Mouse monoclonal antibody raised against Plasmodium vivax MSP1.

Manufacturer:  Abnova Corporation MAB12435

Catalog No. 89-934-592

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

MSP1
6G2
In PBS, pH 7.2 (0.09% sodium azide)
Murine
IgG
1mg
Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA
Unconjugated
Affinity Purified
Native purified Plasmodium vivax MSP1.
Antigen Affinity Purification
RUO
Primary
Parasite
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit