Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSPG, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

169209-64-7
C9H11NO5S
245.249
MVDSFPIEJILRME-UHFFFAOYSA-N
(RS)-α-Methyl-4-sulfonophenylglycine
3964410

Group II/group III mGlu antagonist

Relatively non-selective antagonist of presynaptic mGlu receptors in neonatal rat spinal cord and adult rat cerebrocortical mGlu receptors.
SMILES CC(C1=CC=C(C=C1)S(=O)(=O)O)(C(=O)O)N
IUPAC Name 2-amino-2-(4-sulfophenyl)propanoic acid
Formula Weight 245.25
Quantity 50mg
Chemical Name or Material MSPG
Grade >98%

Recommended Storage : Store at Room Temperature