Learn More
MSRB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 176291AP
Catalog No. 50-173-1694
Add to cart
Description
Description
MSRB2 catalyzes the reduction of free and protein-bound methionine sulfoxide to methionine.
Specifications
Specifications
MSRB2
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4FZX5, Q78J03, Q9Y3D2
CBS 1, CBS1, CGI 131, MSRB, MSRB2, PILB
Rabbit
IgG
150 μL
-20°C
Primary
22921, 361286, 76467
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MSRB2
Liquid
Msrb2
MSRB2 Fusion Protein Ag11881
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat