Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MSTO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552221
Description
Immunogen sequence: GLYRDKQLDA AIAWQGKLTT HKEELYPKNP YLQDFLSAEG VLSSDGVWRV KSIPNGKGSS PLPTATTPKP LIPTEASIRV WSDFLRVHLH PRSICMIQKY NHDGEAGRLE AFGQGESVLK EPKYQ Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 70%.
MSTO1 is a protein coding gene involved in the regulation of mitochondrial distribution and morphology. Required for mitochondrial fusion and mitochondrial network formation.
Specifications
MSTO1 | |
Polyclonal | |
Unconjugated | |
MSTO1 | |
LST005; misato 1, mitochondrial distribution and morphology regulator; misato homolog 1; MST; MSTO1; Protein misato homolog 1; SLTP005 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BUK6 | |
MSTO1 | |
Recombinant protein corresponding to Human MSTO1. Recombinant protein control fragment (Product #RP-89151). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction