Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSTO1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259112

Catalog No. NBP259112


Add to cart

Description

Description

MSTO1 Polyclonal antibody specifically detects MSTO1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

MSTO1
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
DKFZp686B1757, DKFZp686I01261, FLJ10504, LST005, misato homolog 1 (Drosophila), MST, protein misato homolog 1, RP11-29H23.3
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Immunoprecipitation
Immunoprecipitation
Affinity Purified
MSTO1
Maps to a region between 175 and 225
Immunogen affinity purified
RUO
Primary
55154
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only