Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MSX1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5045SP
This item is not returnable.
View return policy
Description
MSX1 Monoclonal specifically detects MSX1 in Human samples. It is validated for Immunohistochemistry.Specifications
MSX1 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
P28360 | |
MSX1 | |
E. coli-derived recombinant human MSX1 Met1-Thr165 Accession # P28360 | |
25 μg | |
Primary | |
Detects human MSX1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human MSX2 is observed. | |
Human | |
Purified |
Immunohistochemistry | |
801601 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Homeobox protein Hox-7, homeobox protein MSX-1, HOX7, HOX7homeobox 7, HYD1, HYD1msh homeobox homolog 1 (Drosophila), msh (Drosophila) homeo box homolog 1 (formerly homeo box 7), msh homeo box 1, msh homeobox 1, Msh homeobox 1-like protein, msh homeobox homolog 1, OFC5, STHAG1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4487 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction