Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MT-ND1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575179
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7), while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 1 (ND1) binds rotenone and rotenone analogs and might be involved in electron transfer to ubiquinone. Mutations in the ND1 gene may be implicated in several disorders, including Leber hereditary optic neuropathy, Alzheimer's disease and Parkinson disease.
Specifications
MT-ND1 | |
Polyclonal | |
Unconjugated | |
mt-Nd1 | |
mitochondrially encoded NADH dehydrogenase 1; Mtnd1; MT-ND1; NADH dehydrogenase; NADH dehydrogenase subunit 1; NADH1; NADH-ubiquinone oxidoreductase chain 1; Nd1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17716, 26193, 4535 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P03886, P03888, P03889 | |
mt-Nd1 | |
Synthetic peptide corresponding to amino acids 189-202 of Mouse MT-ND1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction