Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MT-ND3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5120601

This item is not returnable.
View return policy
Description
Positive test controls include: 293T, PC-3, Mouse heart, Mouse brain.
MT-ND3 Polyclonal antibody specifically detects MT-ND3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
MT-ND3 | |
Polyclonal | |
Unconjugated | |
MT-ND3 | |
1.6.5.3; mitochondrially encoded NADH dehydrogenase 3; Mtnd3; mt-Nd3; NADH dehydrogenase subunit 3; NADH3; NADH-ubiquinone oxidoreductase chain 3; ND3 | |
Recombinant protein of human MT-ND3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
2.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P03897, P03899, P05506 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17718, 26199, 4537 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title