Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT-ND6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550710

Catalog No. PA550710


Add to cart

Description

Description

The antibody detects endogenous levels of total MT-ND6 protein.

The exact function of ND6 remains unknown.
Specifications

Specifications

MT-ND6
Polyclonal
Unconjugated
MT-ND6
Liquid
MT-ND6
Synthetic peptide corresponding to a region derived from internal residues of human mitochondrially encoded NADH dehydrogenase 6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
2.4 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P03923
MTND6, ND6
Rabbit
IgG
100 μL
-20°C
Primary
4541
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit