Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MT-SP1/Matriptase Rabbit anti-Human, Clone: A11 (Azide Free), MilliporeSigma™
Rabbit Monoclonal Antibody
$177.00 - $444.00
Specifications
Antigen | MT-SP1/Matriptase |
---|---|
Clone | A11 (Azide Free) |
Applications | Immunohistochemistry (Paraffin), Inhibition Assays |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC115I25
|
MilliporeSigma
MABC115I25UG |
25 μg |
Each for $177.00
|
|
|||||
MABC115I100
|
MilliporeSigma
MABC115I100UG |
100 μg |
Each for $444.00
|
|
|||||
Description
MT-SP1/Matriptase Monoclonal antibody specifically detects MT-SP1/Matriptase in Human samples. It is validated for Immunohistochemistry (Paraffin), Inhibition.Specifications
MT-SP1/Matriptase | |
Immunohistochemistry (Paraffin), Inhibition Assays | |
Unconjugated | |
Rabbit | |
Human | |
Catalytic residues in the protease catalytic domain of Human MT-SP1/Matriptase. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. |
A11 (Azide Free) | |
Monoclonal | |
Purified | |
RUO | |
Purified rabbit recombinant monoclonal antibody in PBS without preservatives. | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title