Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTCH2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP312914
This item is not returnable.
View return policy
Description
MTCH2 Polyclonal antibody specifically detects MTCH2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MTCH2 | |
| Polyclonal | |
| PBS, 20% Glycerol (pH7) | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 2310034D24Rik, Met-induced mitochondrial protein, MIMP, mitochondrial carrier 2, mitochondrial carrier homolog 2, mitochondrial carrier homolog 2 (C. elegans) | |
| Recombinant protein encompassing a sequence within the center region of human MTCH2. The exact sequence is proprietary. | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 23788 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction