Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTCO2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA588146
Description
Immunogen sequence: MGHQWYWSYE YTDYEDLCFD SYMIPTNDLK PGELRLLEVD NRVVLPMELP IRMLISSEDV LHSWAVPSLG LKTDAIPGRL NQATVTSNRP GLFYGQCSEI C; Positive Samples: Mouse liver, Mouse heart, Mouse kidney, Rat liver, Rat kidney; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein
Cytochrome c oxidase is the fourth complex in the respiratory chain and is responsible for catalyzing the conversion of O2 to H2O. Subunit 2 of the cytochrome c oxidase complex combines with two other subunits (1 and 3) to form a core protein structure that performs many functions of the enzyme. The metallic copper center of this subunit transfers electrons to the heme center of subunit 1, which results in the movement of electrons from cytochrome c to the heme A3 and copper B metallic center of complex IV. Defects in subunit 2 of cytochrome c oxidase can result in COX deficiency, which causes a wide range of symptoms from local myopathy to multiple system pathologies that begin between infancy and adulthood. Abnormalities in this subunit are also associated with tumor development.Specifications
MTCO2 | |
Polyclonal | |
Unconjugated | |
MT-CO2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17709, 26198, 4513 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
4.71 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COX2, MT-CO2 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of mouse MT-CO2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title