Learn More
Tocris Bioscience™ MTEP hydrochloride
Manufacturer:  Tocris Bioscience™ 2921 /50
 View more versions of this product
Catalog No. 29-215-0
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Potent, selective and non-competitive mGlu5 antagonist (IC50 = 5nM in Ca2+-flux assay; Ki= 16nM). Displays anxiolytic activity in vivo and is orally active.
Specifications
Specifications
3-((2-Methyl-1,3-thiazol-4-yl)ethynyl)pyridine hydrochloride
>99%
Observed MW: 236.72
Soluble to 100mM in DMSO
1186195-60-7
C11H8N2S.HCl
50mg
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C