Learn More
Tocris Bioscience™ MTEP hydrochloride Available on GSA/VA Contract for Federal Government customers only.
Potent and selective mGlu5 antagonist
$169.60
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
29-211-0R Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
2921/10
Each for $169.60
Add to cart
29-215-0 View Documents Tocris Bioscience™
2921 /50
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Potent, selective and non-competitive mGlu5 antagonist (IC50 = 5 nM in Ca2+-flux assay; Ki= 16 nM). Displays anxiolytic activity in vivo and is orally active.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.