Learn More
MTF1 Mouse anti-Human, Clone: OTI1D3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF805437
Catalog No. 50-166-7426
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a transcription factor that induces expression of metallothioneins and other genes involved in metal homeostasis in response to heavy metals such as cadmium, zinc, copper, and silver. The protein is a nucleocytoplasmic shuttling protein that accumulates in the nucleus upon heavy metal exposure and binds to promoters containing a metal-responsive element (MRE).
Specifications
Specifications
MTF1
Monoclonal
Unconjugated
MTF1
Lyophilized
MTF1
Human recombit protein fragment corresponding to amino acids 1-272 of human MTF1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1D3
PBS with 8% trehalose and no preservative; pH 7.3
Q14872
MTF-1, ZRF
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4520