Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MTFMT Mouse anti-Human, Clone: OTI1C1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA503547
Description
The protein encoded by this nuclear gene localizes to the mitochondrion, where it catalyzes the formylation of methionyl-tRNA.Specifications
MTFMT | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96DP5 | |
COXPD15, FMT1 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
123263 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C1 | |
Unconjugated | |
MTFMT | |
Liquid | |
MTFMT | |
Human recombit protein fragment corresponding to amino acids 175-389 of human MTFMT produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title